3-Amino-2-pyridone ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone

A2285-1G A2285-5G
TCA2285-1GEA 165.9 CAD
TCA2285-1G TCA2285-5G
3-Amino-2-pyridone ≥96.0% (by GC, titration analysis)
3-Amino-2-pyridone

CAS Number: 33630-99-8
MDL Number: MFCD03411556
Molecular Formula: C5H6N2O
Molecular Weight: 110.12
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: 3-Amino-2-pyridinone, 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol

Formula: C₅H₆N₂O
MW: 110.12 g/mol
Melting Pt: 124 °C
MDL Number: MFCD03411556
CAS Number: 33630-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00acc410b715891a93138653edf8e0ca44c0ff01&vid=2f8beffe7b79f1eb2ac9ae3a8f9162084cb157f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR