N-Acetyl-4-hydroxy-L-proline (cis and trans mixture) ≥97.0%

Supplier: TCI America

Synonyms: N-Acetyl-4-hydroxypyrrolidine-2-carboxylic Acid, Ac-Hyp-OH

A2265-25G A2265-5G
TCA2265-25GEA 219.62 CAD
TCA2265-25G TCA2265-5G
N-Acetyl-4-hydroxy-L-proline (cis and trans mixture) ≥97.0%
N-Acetyl-4-hydroxy-L-proline (cis and trans mixture)

CAS Number: 926905-71-7
Molecular Formula: C7H11NO4
Molecular Weight: 173.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 131
Specific rotation [a]20/D: -117 deg (C=4, H2O)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Acetyl-4-hydroxypyrrolidine-2-carboxylic Acid, Ac-Hyp-OH

Formula: C₇H₁₁NO₄
MW: 173.17 g/mol
Melting Pt: 131 °C
CAS Number: 926905-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17aedaa9d6ea41d9ef98adb2827f2c566fe85589&vid=17aedaa9d6ea41d9ef98adb2827f2c566fe85589&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR