1-Acetylpiperidine-4-carboxylic acid 98,0

Supplier: TCI America

A2261-25G A2261-5G
TCA2261-25GEA 213.3 CAD
TCA2261-25G TCA2261-5G
1-Acetylpiperidine-4-carboxylic acid 98,0
1-Acetylpiperidine-4-carboxylic acid

CAS Number: 25503-90-6
MDL Number: MFCD00023706
Molecular Formula: C8H13NO3
Molecular Weight: 171.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 184


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Acetylisonipecotic Acid

Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00023706
CAS Number: 25503-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dac3b19aed342c0e234bb556ae546a4356cd45b&vid=5dac3b19aed342c0e234bb556ae546a4356cd45b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR