3-Amino-2-naphthoic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

A2258-1G A2258-5G
TCA2258-1GEA 69.52 CAD
TCA2258-1G TCA2258-5G
3-Amino-2-naphthoic acid ≥97.0% (by titrimetric analysis)
3-Amino-2-naphthoic acid

CAS Number: 5959-52-4
MDL Number: MFCD00004115
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 212

Formula: C₁₁H₉NO₂
Melting Pt: 212 °C
MDL Number: MFCD00004115
CAS Number: 5959-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=300482983bd055ecab30f75d72f288423eda76bf&vid=300482983bd055ecab30f75d72f288423eda76bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR