3-Anilino-1-propanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2248-5G A2248-25G
TCA2248-5GEA 223.36 CAD
TCA2248-5G TCA2248-25G
3-Anilino-1-propanol ≥98.0% (by GC, titration analysis)
3-Anilino-1-propanol

CAS Number: 31121-11-6
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Specific Gravity (20/20): 1.08
Lambda max.: 245 nm (Hexane)Storage Temperature: 0-10°C


Other Product Names / Synonyms: N-(3-Hydroxypropyl)aniline

Formula: C₉H₁₃NO
Boiling Pt: 140 °C
Density: 1.08 (20 °C)
MDL Number: MFCD15206074
CAS Number: 31121-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1924b921853c8cdb1cdd726397ef955548bcbf12&vid=1924b921853c8cdb1cdd726397ef955548bcbf12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR