2-Amino-6-chloro-4-nitrophenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A2245-25G A2245-5G
TCA2245-25GEA 494.54 CAD
TCA2245-25G TCA2245-5G
2-Amino-6-chloro-4-nitrophenol ≥98.0% (by titrimetric analysis)
2-Amino-6-chloro-4-nitrophenol

CAS Number: 6358-09-4
MDL Number: MFCD00035767
Molecular Formula: C6H5ClN2O3
Molecular Weight: 188.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 162


Other Product Names / Synonyms: 3-Chloro-2-hydroxy-5-nitroaniline

Formula: C₆H₅ClN₂O₃
Melting Pt: 162 °C
MDL Number: MFCD00035767
CAS Number: 6358-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ce69de87be7438da2baac7d659d28c6d4776bc8&vid=1ce69de87be7438da2baac7d659d28c6d4776bc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR