4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine ≥97.0%
Supplier: TCI America
Synonyms:
4-Amino-N-Boc-D-phenylalanine, Boc-D-Phe(4-NH₂)-OH, Boc-D-Phe(4-NH2)-OH
A2130-1G
A2130-5G
TCA2130-1GEA
109.02
CAD
TCA2130-1G
TCA2130-5G
4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine ≥97.0%
4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine
CAS Number: 164332-89-2
MDL Number: MFCD00236818
Molecular Formula: C14H20N2O4
Molecular Weight: 280.32
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -25 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Amino-N-Boc-D-phenylalanine, Boc-D-Phe(4-NH2)-OH
Formula:
C₁₄H₂₀N₂O₄ MW: 280.32 g/mol |
MDL Number:
MFCD00236818 CAS Number: 164332-89-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...