4-Amino-2,5-xylenol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

A2116-25G A2116-5G
TCA2116-25GEA 598.35 CAD
TCA2116-25G TCA2116-5G
4-Amino-2,5-xylenol ≥97.0% (by GC, titration analysis)
4-Amino-2,5-xylenol

CAS Number: 3096-71-7
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 238


Other Product Names / Synonyms: 4-Amino-2,5-dimethylphenol, 4-Hydroxy-2,5-dimethylaniline

Formula: C₈H₁₁NO
Melting Pt: 238 °C
MDL Number: MFCD00007873
CAS Number: 3096-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e80e02c1f13555134ed28b99116ce6f591c78d35&vid=e80e02c1f13555134ed28b99116ce6f591c78d35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR