6-Amino-2-naphthoic acid ≥97.0%

Supplier: TCI America

Synonyms: 6-Amino-2-naphthoic acid, 6-Amino-2-naphthalenecarboxylic acid, 6-Amino-2-naphthoic Acid

A1938-25G A1938-5G
TCA1938-25GEA 805.8 CAD
TCA1938-25G TCA1938-5G
6-Amino-2-naphthoic acid ≥97.0%
6-Amino-2-naphthoic acid

CAS Number: 116668-47-4
MDL Number: MFCD01861831
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 225


Other Product Names / Synonyms: 6-Amino-2-naphthalenecarboxylic Acid

Formula: C₁₁H₉NO₂
Melting Pt: 225 °C
MDL Number: MFCD01861831
CAS Number: 116668-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=805d7ff97811d2e994f56f9cb2aa2bd1a21e15e2&vid=805d7ff97811d2e994f56f9cb2aa2bd1a21e15e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR