3-Amino-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

A1859-25G A1859-5G
TCA1859-25GEA 135 CAD
TCA1859-25G TCA1859-5G
3-Amino-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Amino-1H-pyrazole

CAS Number: 1820-80-0
Molecular Formula: C3H5N3
Molecular Weight: 83.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 282
Flash Point (°C): 110
Freezing point (°C): 35
Storage Temperature: 0-10°C

Formula: C₃H₅N₃
MW: 83.09 g/mol
Boiling Pt: 282 °C
Flash Pt: 110 °C
MDL Number: MFCD00005236
CAS Number: 1820-80-0
UN: 3259
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cf3fcb2ba92b001d1f194f070285cf8357a41ff&vid=3cf3fcb2ba92b001d1f194f070285cf8357a41ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR