5-Amino-2-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine

A1816-1G A1816-5G
TCA1816-1GEA 113.76 CAD
TCA1816-1G TCA1816-5G
5-Amino-2-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
5-Amino-2-(trifluoromethyl)pyridine

CAS Number: 106877-33-2
MDL Number: MFCD01862656
Molecular Formula: C6H5F3N2
Molecular Weight: 162.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₆H₅F₃N₂
MW: 162.11
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD01862656
CAS Number: 106877-33-2
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca596482851f9f6cfdcc1c6d055a4afa19692782&vid=ca596482851f9f6cfdcc1c6d055a4afa19692782&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR