Methyl hydrogen adipate ≥97.0%

Supplier: TCI America

Synonyms: 1,6-Hexanedioic acid methyl ester, Adipic acid methyl ester, Adipic acid monomethyl ester, 1,6-Hexanedioic acid monomethyl ester, Monomethyl adipate

A1738-25G A1738-100G
TCA1738-25GEA 97.49 CAD
TCA1738-25G TCA1738-100G
Methyl hydrogen adipate ≥97.0%
Methyl hydrogen adipate

CAS Number: 627-91-8
MDL Number: MFCD00004418
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 162
Melting point (°C): 9
Flash Point (°C): 110
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Adipic Acid Monomethyl Ester, Methyl Hydrogen Adipate, Monomethyl Hexanedioate, Hexanedioic Acid Monomethyl Ester

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 292
Melting Pt: 9 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004418
CAS Number: 627-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2dfde55707b5f2f5a8c2e4a797da37a1ac6f530&vid=c2dfde55707b5f2f5a8c2e4a797da37a1ac6f530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR