4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1577-25G A1577-5G
TCA1577-25GEA 158 CAD
TCA1577-25G TCA1577-5G
4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)
4-Amino-2-methyl-1-butanol

CAS Number: 44565-27-7
MDL Number: MFCD03093615
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.94

Formula: C₅H₁₃NO
Density: 0.94 (20 °C)
MDL Number: MFCD03093615
CAS Number: 44565-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b76eb98ebbc236576f9eb9c3fdc197d5394d6b7&vid=4b76eb98ebbc236576f9eb9c3fdc197d5394d6b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR