(R)-4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1576-1G A1576-5G
TCA1576-1GEA 47.87 CAD
TCA1576-1G TCA1576-5G
(R)-4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)
(R)-4-Amino-2-methyl-1-butanol

CAS Number: 88390-32-3
MDL Number: MFCD03093614
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 80
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: 14.5 deg (neat)

Formula: C₅H₁₃NO
MW: 103.17 g/mol
Boiling Pt: 80 °C
Density: 0.94 (20 °C)
MDL Number: MFCD03093614
CAS Number: 88390-32-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8011153ace01ddb467325e9e246ebbf9bbe60f09&vid=8011153ace01ddb467325e9e246ebbf9bbe60f09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR