N-Boc-1,6-diaminohexane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-tert-Butoxycarbonyl-1,6-hexanediamine, N-Boc-1,6-Diaminohexane
CAS Number: 51857-17-1
MDL Number: MFCD00671489
Molecular Formula: C11H24N2O2
Molecular Weight: 216.33
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 125
Specific Gravity (20/20): 0.96
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(6-Aminohexyl)carbamic Acid tert-Butyl Ester, N-Boc-1,6-diaminohexane, N-Boc-1,6-hexanediamine, N-(tert-Butoxycarbonyl)-1,6-hexanediamine, tert-Butyl N-(6-Aminohexyl)carbamate
Formula:
C11H24N2O2 MW: 216.33 g/mol Density: 0.96 (20 °C) Flash Pt: 125 °C |
MDL Number:
MFCD00671489 CAS Number: 51857-17-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...