DL-ɑ-Amino-ε-caprolactam 98,0

Supplier: TCI America

Synonyms: DL-3-Amino-2-oxohexamethyleneimine, DL-3-Aminohexahydro-2-azepinone, 3-Amino-2-azepanone(Chunks or pellets or lump), 3-Amino-2-azepanone

A1003-5G A1003-1G
TCA1003-5GEA 157.05 CAD
TCA1003-5G TCA1003-1G
DL-ɑ-Amino-ε-caprolactam 98,0
DL-alpha-Amino epsilon caprolactam

CAS Number: 671-42-1
MDL Number: MFCD00064475
Molecular Formula: C6H12N2O
Molecular Weight: 128.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 172
Melting point (°C): 77
Flash Point (°C): 173


Other Product Names / Synonyms: DL-3-Aminohexahydro-2-azepinone, DL-3-Amino-2-oxohexamethyleneimine

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
Boiling Pt: 172 °C
Melting Pt: 77 °C
Flash Pt: 173 °C
MDL Number: MFCD00064475
CAS Number: 671-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=193ea0c6da6d1e05c995c6517a941922eabaae0e&vid=193ea0c6da6d1e05c995c6517a941922eabaae0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR