2-Aminofluorene ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0621-25G A0621-5G
TCA0621-25GEA 246.32 CAD
TCA0621-25G TCA0621-5G
2-Aminofluorene ≥98.0% (by HPLC, titration analysis)
2-Aminofluorene

CAS Number: 153-78-6
MDL Number: MFCD00001125
Molecular Formula: C13H11N
Molecular Weight: 181.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 127


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 2-Fluorenamine

Formula: C₁₃H₁₁N
Melting Pt: 127 °C
MDL Number: MFCD00001125
CAS Number: 153-78-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d61875f97a9aeb6da411624ea503ae13c98d10b4&vid=d61875f97a9aeb6da411624ea503ae13c98d10b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR