9-Aminoacridine hydrochloride monohydrate ≥98.0%

Supplier: TCI America

Synonyms: 9-Aminoacridine hydrochloride hydrate, Acridin-9-amine hydrochloride hydrate

A0619-25G A0619-5G
TCA0619-25GEA 110.6 CAD
TCA0619-25G TCA0619-5G
9-Aminoacridine hydrochloride monohydrate ≥98.0%
9-Aminoacridine hydrochloride monohydrate

CAS Number: 52417-22-8
MDL Number: MFCD00150071
Molecular Formula: C13H10N2
Molecular Weight: 230.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₃H₁₁ClN₂·1H₂O
MW: 248.71 g/mol
MDL Number: MFCD00150071
CAS Number: 52417-22-8
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19a0780aa8ad5d64cdbb90fadf8d2c659a9fe0ca&vid=19a0780aa8ad5d64cdbb90fadf8d2c659a9fe0ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR