AMP (2-Amino-2-methylpropanol) ≥93.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP

A0333-25ML A0333-500ML
TCA0333-25MLEA 56.88 CAD
TCA0333-25ML TCA0333-500ML
AMP (2-Amino-2-methylpropanol) ≥93.0% (by GC, titration analysis)
AMP (2-Amino-2-methylpropanol)

CAS Number: 124-68-5
MDL Number: MFCD00008051
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >93.0% (GC,T)
Form: Crystal
Boiling point (°C): 165
Flash Point (°C): 75
Freezing point (°C): 23


Other Product Names / Synonyms: AMP

Formula: (CH₃)₂C(NH₂)CH₂OH
MW: 89.14 g/mol
Boiling Pt: 165 °C
Flash Pt: 75 °C
MDL Number: MFCD00008051
CAS Number: 124-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a97bee4896391e0e2d0b3d180e39e84cff2f523b&vid=a97bee4896391e0e2d0b3d180e39e84cff2f523b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR