Dopamine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1-(2-Aminoethyl)-3,4-dihydroxybenzene HCl, 3-Hydroxytyraminium chloride, 3-Hydroxytyramine hydrochloride, 3,4-Dihydroxyphenethylammonium chloride

A0305-1G A0305-25G A0305-5G
TCA0305-1GEA 30.56 CAD
TCA0305-1G TCA0305-25G TCA0305-5G
Dopamine hydrochloride ≥98.0%
Dopamine hydrochloride

CAS Number: 62-31-7
MDL Number: MFCD00012898
Molecular Formula: C8H11NO2
Molecular Weight: 189.64
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: 4-(2-Aminoethyl)pyrocatechol Hydrochloride, 2-(3,4-Dihydroxyphenyl)ethylamine Hydrochloride, Dopamine Hydrochloride

Formula: C₈H₁₂ClNO₂
MW: 189.64
Density: 1.356
MDL Number: MFCD00012898
CAS Number: 62-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92a8a471fb2c6c97c04f7194d7149045ee707e4b&vid=92a8a471fb2c6c97c04f7194d7149045ee707e4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR