Rat Recombinant EGF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
300-05-100UG 300-05-1MG 300-05-20UG 300-05-500UG 300-05-250UG
10788-392EA 354.2 CAD
10788-392 10788-394 10788-788 10821-748 10821-928
Rat Recombinant EGF (from E. coli)
Proteins and Peptides
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.