Rat Recombinant GM-CSF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
300-03-100UG 300-03-1MG 300-03-20UG 300-03-5UG 300-03-500UG
10788-378EA 884.72 CAD
10788-378 10788-380 10788-382 10788-384 10822-030
Rat Recombinant GM-CSF (from E. coli)
Proteins and Peptides
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF promotes immune system development and regulates neutrophil function during infection.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.