Rat Recombinant FGF-basic (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
300-02-10UG 300-02-1MG 300-02-50UG 300-02-500UG 300-02-100UG
10788-366EA 141.54 CAD
10788-366 10788-368 10788-370 10821-756 10821-964
Rat Recombinant FGF-basic (from E. coli)
Proteins and Peptides
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic binds heparin in order to signal through fibroblast growth factor (FGF) receptor tyrosine kinases.
  • High Quality
  • Low Endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Caution: For research use only.