Mouse Recombinant IL-17F (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-62-100UG 200-62-1MG 200-62-25UG 200-62-5UG 200-62-500UG
10788-288EA 884.72 CAD
10788-288 10788-290 10788-292 10788-294 10822-180
Mouse Recombinant IL-17F (from E. coli)
Proteins and Peptides
Interleukin 17F (IL-17F), a member of the IL-17 cytokine family, is secreted by activated CD4+ T cells and monocytes. IL-17F binds the IL-17 receptor related molecule, IL17RC, to promote the production of the interleukin 6 (IL-6), interleukin 8 (IL-8), and granulocyte macrophage colony-stimulating factor (GM-CSF) cytokines. IL-17F also functions to regulate matrix turnover rates, inhibit endothelial cell angiogenesis, and induce the endothelial cell expression of interleukin 2 (IL-2), monocyte chemoattractant protein-1 (MCP-1), and transforming growth factor beta 1 (TGF-β1).
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.