Mouse Recombinant IL-17A (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-59-1MG 200-59-25UG 200-59-5UG 200-59-500UG 200-59-100UG
10788-266EA 6375.71 CAD
10788-266 10788-268 10788-270 10822-160 77660-844
Mouse Recombinant IL-17A (Animal free) (from E. coli)
Proteins and Peptides
Interleukin 17A (IL-17A), also known as CTLA-8, is a member of the IL-17 family of proteins. IL-17A is a proinflammatory cytokine that is secreted by activated CD4+ and CD8+ T lymphocytes. IL-17A acts through its receptor, IL-17R, to promote increased cytokine and chemokine secretion. In turn, the cytokines and chemokines mediate the immunoregulatory function of IL-17A by promoting the proliferation, maturation, and chemoattraction of neutrophils to inflammatory sites.  Elevated levels of IL-17A are associated with rheumatoid arthritis, airway inflammation, allograft rejection, inflammatory bowel disease, psoriasis, cancer, and multiple sclerosis.  Human, mouse, and rat IL-17A show activity on mouse cells.  
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.