Mouse Recombinant PDGF-BB (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

200-58-100UG 200-58-10UG 200-58-1MG 200-58-500UG
10788-258EA 1001.15 CAD
10788-258 10788-260 10788-262 10788-264 10821-872
Mouse Recombinant PDGF-BB (from E. coli)
Proteins and Peptides

Interleukin 17AF (IL-17AF) is a heterodimer that is composed of the interleukin 17A (IL-17A) and interleukin 17F (IL-17F) members of the IL-17 family of cytokines. IL-17AF is produced by T helper 17 cells (Th17) following interleukin 23 (IL-23) stimulation.


  • High quality
  • Low endotoxin
  • Affordable


IL-17AF signals through the IL-17RA/IL-17RC receptor complex and functions to regulate inflammatory responses. IL-17AF induces chemokine and airway neutrophilia production, similar in function to IL-17A and IL-17F homodimers. In regard to these functions, IL-17AF is less active than the IL-17A homodimer and shows greater activity than the IL-17F homodimer. Human and rat IL-17AF are active on mouse cells.


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name PDGF-BB
Environmentally Preferable
Protein synonyms ODGF|GDGF
Protein/peptide type Recombinant
Protein Function Cytokine
Species Mouse
Source E. coli
Conjugation Unconjugated
Gene ID P31240
Sequence MSLGSLAAAE PAVIAECKTR TEVFQISRNL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRASQVQMRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETIVTPRPVT
Protease-free N
Animal-free N
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤1
UniProtKB P31240
Biological activity Bioactivity assay: 3T3 cell proliferation, ED50 ≤2 ng/ml, bioactivity ≥5.0×10⁵ units/mg
Purity ≥95%
Grade RUO
Formulation 10 mM sodium citrate, pH 3.0
Molecular weight 12.4/24.7 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml
Storage conditions −20 °C
Shipping temperature RT
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR