Mouse Recombinant EBI3 Sub-unit (IL-27/IL-35) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Epstein-Barr virus induced gene 3 (EBI3) is a secreted glycoprotein belonging to the hematopoietin receptor family related to the p40 subunit of interleukin 12 (IL-12). EBI3 expression is induced in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form interleukin 27 (IL-27), and with p35 to form interleukin 35 (IL-35). Both IL-27 and IL-35 have anti-inflammatory and regulatory activity.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.