Rat Recombinant IL-1 B (from E. coli)
Supplier: Fujifilm Irvine Scientific
300-30-100UG 300-30-10UG 300-30-1MG 300-30-500UG
10788-516EA 1368.83 CAD
10788-516 10788-518 10788-520 10788-522 10822-080
Rat Recombinant IL-1 B (from E. coli)
Proteins and Peptides
Interleukin 1 beta (IL-1β) is a pro-inflammatory cytokine that is produced by monocytes, macrophages, and dendritic cells (DCs). IL-1β signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter. IL-1β and the independently regulated IL-1α protein have overlapping proinflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.