Mouse Recombinant PDGF-AA (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-54-100UG 200-54-10UG 200-54-1MG 200-54-500UG
10788-234EA 1001.15 CAD
10788-234 10788-236 10788-238 10788-240 10822-334
Mouse Recombinant PDGF-AA (from E. coli)
Proteins and Peptides
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis.  PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β, and EGF signaling.  PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.
  • High quality
  • Low endotoxin
  • Affordable
PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB).  The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
Caution: For research use only.