Mouse Recombinant EGF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-53-100UG
200-53-1MG
200-53-500UG
200-53-5MG
10788-222EA
143.42
CAD
10788-222
10788-224
10788-226
10821-744
Mouse Recombinant EGF (from E. coli)
Proteins and Peptides
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR).
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.