Mouse Recombinant VEGF-120 (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

200-52-10UG 200-52-1MG 200-52-500UG 200-52-100UG
10788-216EA 325.48 CAD
10788-216 10788-218 10788-220 10822-416 77660-842
Mouse Recombinant VEGF-120 (from E. coli)
Proteins and Peptides

Vascular endothelial growth factor A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways. Mouse VEGF-A has several alternatively spliced isoforms, with one being VEGF-120. VEGF-120 is an angiogenic factor that is expressed throughout endochondral bone development and is important during skeletogenesis.


  • High Quality
  • Low Endotoxin
  • Affordable


A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  


Caution: For research use only.

New Product

Order Now

SPECIFICATIONS

Protein/peptide name VEGF-120 VEGF-120
Environmentally Preferable
Protein synonyms glioma-derived endothelial cell mitogen|VEGF-A|VPF glioma-derived endothelial cell mitogen|VEGF-A|VPF
Protein/peptide type Recombinant Recombinant
Protein Function Cytokine Cytokine
Species Mouse Mouse
Source E. coli E. coli
Conjugation Unconjugated Unconjugated
Gene ID Q00731-3 Q00731-3
Sequence MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R
Protease-free N N
Animal-free N N
Nuclease-free N N
Carrier-free Y Y
Endotoxin-free N N
Endotoxin level Low Low
Endotoxin content Endotoxin LAL (EU/µg) ≤0.1 Endotoxin LAL (EU/µg) ≤0.1
Biological activity Bioactivity assay: HUVEC proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2.0×10⁵ units/mg Bioactivity assay: HUVEC proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2.0×10⁵ units/mg
Purity ≥95% ≥95%
Grade RUO
Formulation 0.1% Trifluoroacetic Acid (TFA) 0.1% Trifluoroacetic Acid (TFA)
Molecular weight 14.2/28.4 kDa 14.2/28.4 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml Sterile water at 0.1 mg/ml
Storage conditions –20 °C –20 °C
Shipping temperature RT RT
Tested applications Bioassay Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR