Mouse Recombinant IL-16 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-49-100UG 200-49-10UG 200-49-1MG 200-49-500UG
10788-184EA 1310.06 CAD
10788-184 10788-186 10788-188 10788-190 10822-156
Mouse Recombinant IL-16 (from E. coli)
Proteins and Peptides
Interleukin 16 (IL-16) is produced by CD4+ and CD8+ T cells and functions as a chemoattractant for lymphocytes, monocytes, eosinophils, dendritic cells, and Langerhans cells.  Additionally, IL-16 upregulates the expression of the interleukin 2 receptor (CD25), modulates T lymphocyte growth, and suppresses the replication of the human immunodeficiency virus (HIV) and simian immunodeficiency viruses (SIV).
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.