Human Recombinant Eotaxin (from E. coli)

Supplier: FUJIFILM IRVINE SCIENTIFIC, INC

100-178-5UG 100-178-20UG 100-178-100UG 100-178-1MG 100-178-500UG
10788-548EA 141.54 CAD
10788-548 10788-550 10788-552 10788-554 10821-934
Human Recombinant Eotaxin (from E. coli)
Proteins and Peptides

Eotaxin, also known as Eotaxin-1 and CCL11, is a chemokine that recruits eosinophils to sites of allergic inflammation. Eotaxin signals through the G protein-coupled chemokine receptors CCR2, CCR3, and CCR5. Eotaxin also negatively regulates adult neurogenesis, and high Eotaxin levels are associated with impaired learning and memory.


  • High Quality
  • Low Endotoxin
  • Affordable


A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  


Caution: For research use only.

Order Now

SPECIFICATIONS

Protein/peptide name Eotaxin
Protein synonyms Eosinophil chemotactic protein|CCL11
Environmentally Preferable
Protein/peptide type Recombinant
Protein Function Cytokine
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID P51671
Sequence GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP
Protease-free N
Animal-free N
Nuclease-free N
Carrier-free Y
Endotoxin-free N
Endotoxin level Low
Endotoxin content Endotoxin LAL (EU/µg) ≤0.1
Biological activity Bioactivity: No biological activity data is available at this time
Purity ≥95%
Grade RUO
Formulation 0.1% Trifluoroacetic Acid (TFA)
Molecular weight 8.4 kDa
Reconstitution instructions Sterile water at 0.1 mg/ml
Storage conditions –20 °C
Shipping temperature RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR