Human Recombinant Eotaxin-3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-179-5UG
100-179-20UG
100-179-100UG
100-179-1MG
100-179-500UG
10788-556EA
141.54
CAD
10788-556
10788-558
10788-560
10788-562
10821-936
Human Recombinant Eotaxin-3 (from E. coli)
Proteins and Peptides
Eotaxin-3, also known as CCL26, MIP-4-alpha, and TSC-1, is a chemokine that is made by vascular endothelial and lung epithelial cells following interleukin 4 (IL-4) or interleukin 13 (IL-13) stimulation. Eotaxin-3 signals through the G protein-coupled chemokine receptor CCR3 to recruit eosinophils and basophils to inflammatory sites.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.