Mouse Recombinant MIP-1beta / CCL4 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Macrophage inflammatory protein-1 beta (MIP-1 β), also known as CCL4, is produced by macrophages and functions as a mitogen-inducible cytokine. MIP-1 β signals through the chemokine receptor CCR5 to chemoattract immune cells. MIP-1 β induces inflammatory responses, including neutrophil superoxide production. The MIP-1 β and MIP-1 α heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1).
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.