Mouse Recombinant MCP-1 / CCL2 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-39-100UG 200-39-10UG 200-39-1MG 200-39-500UG
10788-130EA 884.72 CAD
10788-130 10788-132 10788-134 10788-136 10822-248
Mouse Recombinant MCP-1 / CCL2 (from E. coli)
Proteins and Peptides
Monocyte chemotactic protein 1 (MCP-1), also known as CCL2, is produced by injured or infected tissues. MCP-1 signals through the CCR2 and CCR4 G protein-coupled receptors to recruit memory T cells, monocytes, and dendritic cells to sites of inflammation.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.