Mouse Recombinant GDNF (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
200-37-100UG
200-37-10UG
200-37-1MG
200-37-500UG
10788-106EA
1365.33
CAD
10788-106
10788-108
10788-110
10788-112
10822-014
Mouse Recombinant GDNF (from E. coli)
Proteins and Peptides
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.