Human Recombinant Oncostatin M (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-56-10UG 100-56-100UG 100-56-1MG 100-56-500UG
10789-112EA 147.52 CAD
10789-112 10789-114 10789-116 10789-118 10822-326
Human Recombinant Oncostatin M (from E. coli)
Proteins and Peptides
Oncostatin M is a cytokine that is produced by macrophages, dendritic cells, and T lymphocytes during inflammatory events. The Type-I and Type-II Oncostatin M receptors are located on the cell surface of endothelial and tumor cells, contain the glycoprotein 130 (gp130) subunit, and activate the JAK/STAT signaling pathway. Oncostatin M functions to inhibit tumor cell proliferation, induce liver stem cell maturation, regulate cytokine production during hematopoiesis and inflammation, stimulate bone formation, and promote nervous system development.
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Caution: For research use only.