AtmoSense™ Moisture Monitor
Supplier: Labconco
5255600 5255690
10002-754EA 7394.4 CAD
10002-754 77574-086
AtmoSense™ Moisture Monitor
Fume Hoods and Enclosures Chemical Fume Hoods
The AtmoSense™ moisture monitor is designed for general purpose trace moisture detection as low as 0 to 20 ppm.
Designed for placement atop transfer chamber of a Labconco® Protector® controlled atmosphere, combination glove box, or the Precise® controlled atmosphere glove box rather than inside. Operates in-line with the AtmosPure™ Re-Gen gas purifier. Constructed with a durable glacier white powder-coated housing. Gas inlet and outlet connections are 1.9 cm (³/₄") brass compression fittings.
An analog output signal of 0 to 5 volts is provided from –50 to 10 °C dew point (64 to 12,310 ppm). Auto-calibration occurs every 30 minutes and is integrated between 20 to 1,250 ppm.
The device provides a digital LCD readout from 0 to 12,310 ppm moisture (0 to 52% RH), with resolution accuracy of 5 ppm (2 °C dew point) measured at 20 ppm (–60 °C dew point). Includes certificate of initial calibration.
Ordering information: The AtmoSense™ moisture monitor must be used with circulation tubing kit 5441808 (sold separately). The AtmoSense™ moisture monitor includes 100 to 115 volt cord with IEC 320 C14 plug for North America/Japan use. With 12 VDC input voltage for operation on 50/60Hz powered from 100, 115, or 208/240 volt AC. Power cords with other plug types are available (sold separately).