Metal Slide Storage Cabinets and Bases, Globe Scientific
Supplier: Globe Scientific
513500T 513500W 513503G 513503T 513503W 513500G
10014-740EA 549.37 CAD
10014-740 10014-742 10014-744 10014-746 10014-748 10014-752
Metal Slide Storage Cabinets and Bases, Globe Scientific
Slide Holders
This sturdy, heavy-duty, high capacity metal slide storage cabinet is an ideal solution for the safe, long-term storage of slide specimens
  • Each cabinet unit contains 6 dual slotted drawers
  • Each drawer holds up to 750 slides (4500 slides per 6 drawer cabinet unit)
  • One base is required for each new stack of (up to) 10 cabinets
  • Interlocking system locks the stacked cabinet units together
  • Each drawer has brass pull knob and identification card holder - identification cards included
  • Included foam blocks keep slides upright in drawers
  • Fits all standard sized microscope slides: 25×75 mm or 3×1"
Each drawer features a brass pull knob and identification card holder (identification cards included). Fitting all standard sized microscope slides (25×75 mm or 3×1"), the drawers also include foam blocks to keep slides upright while in storage.
Each cabinet unit holds up to 4500 slides. Up to 10 units can be secured together on a single base for a total of 45000 slides per stack.