Scout® Balance with Backlit LCD Screen, Ohaus®
Supplier: OHAUS CORP.
30253029 30253030 30253031 30253032 30253033 30253034 30253035 30253036 30253037 30253038 30253039
470217-930EA 1414.7 CAD
470217-930 470217-932 470217-934 470217-936 470217-938 470217-940 470217-942 470217-944 470217-946 470217-948 470217-950
Scout® Balance with Backlit LCD Screen, Ohaus®
Balances and Scales
These Scout balances are the next generation of portable balances for the classroom.
  • Superior Overload Protection
  • Stackable Design for Convenient Storage
  • Bright Backlit LCD and Four-Button Operation
  • Multiple Connectivity Options for Data Logging and Communication
The new Ohaus Scout weighs faster and stores easier, making it the perfect option for science classrooms that require greater versatility, performance, and technological capabilities. Designed specifically for educational weighing applications, the new Ohaus Scout with Backlit LCD offers the same superior overload protection as before with a new, sleek, and stackable design.
The new Scout also has a variety of communication options such as Bluetooth, Ethernet, USB, and more. Combined with a simplified four-button operation, the Scout's large backlit display increases readability in dim lighting conditions and makes it easier to view results from across a desk or lab bench.
Measures in g, kg, N, oz, lb, lb:oz
Ordering information: Five-year manufacturer's warranty is included with purchase.