Anti-LY6C1 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RB6-8C5]

Supplier: Tonbo Biosciences
60-5931-U025 60-5931-U100
10051-528EA 94.8 CAD
10051-528 10051-530
Anti-LY6C1 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RB6-8C5]
Antibodies
Type: Primary
Antigen: LY6C1
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR