Ghost Dye™ Violet 450 for flow cytometry

Supplier: Tonbo Biosciences

13-0863-T100 13-0863-T500
10140-978EA 80.58 CAD
10140-978 10141-462
Ghost Dye™ Violet 450 for flow cytometry
Ghost Dye™

Ghost Dye™ Violet 450 is an amine reactive viability dye that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications.


  • Excitation Laser Violet (405 nm)
  • Emission (nm) 450
  • Formulation 1 µL/test in DMSO


This dye irreversibly binds free amines available on the cell surface as well as intracellular free amines exposed in cells with compromised cell membranes. Necrotic cells with compromised membranes will react with significantly more Ghost Dye™ Violet 450 dye than viable cells in the same sample and therefore will exhibit much greater fluorescence intensity allowing exclusion of these cells from analysis.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR