Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Columbia Biosciences
D5-110-FC-100 D5-110-FC-1MG
10065-880EA 262.39 CAD
10065-880 10065-882
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: F(c) gamma chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR