Anti-IgG Goat Polyclonal Antibody (Europium 1024)

Supplier: Columbia Biosciences
D17-112-1MG D17-112-50 D17-112-20
10065-532EA 15995.37 CAD
10065-532 10065-534 10065-536
Anti-IgG Goat Polyclonal Antibody (Europium 1024)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR