Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: Columbia Biosciences
D3-114-100 D3-114-1MG
10065-614EA 273.53 CAD
10065-614 10065-616
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR