Anti-DYKDDDDK Mouse Polyclonal Antibody (DyLight® 650) [clone: M2]

Supplier: Columbia Biosciences
D10-1718-100 D10-1718-1MG
10065-486EA 890.5 CAD
10065-486 10065-488
Anti-DYKDDDDK Mouse Polyclonal Antibody (DyLight® 650) [clone: M2]
Antibodies
mouse monoclonal [M2] to DDDDK tag (binds to FLAG tag sequence (Dylight 650)

Type:
Antigen: DYKDDDDK
Clonality: Polyclonal
Clone: M2
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR