Anti-EQKLISEEDL Mouse Monoclonal Antibody (Europium 1024) [clone: 9E10]

Supplier: Columbia Biosciences
D17-1714-1MG D17-1714-50 D17-1714-20
10065-556EA 25216.09 CAD
10065-556 10065-558 10065-560
Anti-EQKLISEEDL Mouse Monoclonal Antibody (Europium 1024) [clone: 9E10]
Antibodies
Type:
Antigen: EQKLISEEDL
Clonality: Monoclonal
Clone: 9E10
Conjugation: Europium 1024
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: c-myc tagged proteins
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR