Anti-GST Goat Polyclonal Antibody (Europium 1024)

Supplier: Columbia Biosciences
D17-1310-1MG D17-1310-50 D17-1310-20
10065-544EA 18409.54 CAD
10065-544 10065-546 10065-548
Anti-GST Goat Polyclonal Antibody (Europium 1024)
Antibodies
Type:
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR