Anti-DLG4 Mouse Monoclonal Antibody [clone: 7000]

Supplier: Rockland Immunochemical
200-301-G33
RL200-301-G33EA 854.78 CAD
RL200-301-G33
Anti-DLG4 Mouse Monoclonal Antibody [clone: 7000]
Antibodies
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone: 7000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Bovine, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR